siRNA Details
| Gene Symbol: Slitrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107587.1 | NP_001101057.1 | 2 | 871 | ||
| XM_006257625.2 | XP_006257687.1 | 3 | 1153 | |||
| XM_006257626.2 | XP_006257688.1 | 4 | 873 | |||
| XM_008773586.1 | XP_008771808.1 | 3 | 1164 | |||

Slitrk2
Genomic Map