siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107596.1 | NP_001101066.1 | 21 | 3849 | ||
| XM_006231163.1 | XP_006231225.1 | 22 | 3898 | |||
| XM_006231164.2 | XP_006231226.1 | 23 | 4172 | |||
| XM_006231165.2 | XP_006231227.1 | 23 | 5010 | |||
| XM_006231166.2 | XP_006231228.1 | 23 | 4006 | |||
| XM_008760298.1 | XP_008758520.1 | 24 | 4347 | |||
| XM_008760299.1 | XP_008758521.1 | 23 | 4058 | |||

Tmem2
Genomic Map