siRNA Details
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_219909.8 | XP_219909.7 | 7 | 963 | ||
GenBank mRNA | BC166504.1 | - | 7 | 748 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_219909.8 | XP_219909.7 | 7 | 963 | ||
GenBank mRNA | BC166504.1 | - | 7 | 748 |