siRNA Details
| Gene Symbol: Fbxw4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107600.1 | NP_001101070.1 | 2, 3 | 509 | ||
| Gene Symbol: LOC100911855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760502.1 | XP_008758724.1 | 2, 3 | 503 | ||

Fbxw4
Genomic Map