siRNA Details
| Gene Symbol: Ifit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007694.1 | NP_001007695.1 | 2 | 1076 | ||
| GenBank mRNA | BC079079.1 | - | 2 | 1076 | 7122607 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifit3| Gene Symbol: Ifit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007694.1 | NP_001007695.1 | 2 | 1076 | ||
| GenBank mRNA | BC079079.1 | - | 2 | 1076 | 7122607 | |