siRNA Details
| Gene Symbol: Rbm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107611.2 | NP_001101081.2 | 11 | 3332 | ||
| GenBank mRNA | EU562301.1 | - | 11 | 3332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm20| Gene Symbol: Rbm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107611.2 | NP_001101081.2 | 11 | 3332 | ||
| GenBank mRNA | EU562301.1 | - | 11 | 3332 | ||