siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748666.3 | XP_003748714.1 | 7 | 1040 | ||
| XM_006227693.2 | XP_006227755.1 | 6 | 2696 | |||
| XM_006227695.2 | XP_006227757.1 | 7 | 754 | |||
| XM_006227696.2 | XP_006227758.1 | 6 | 673 | |||
| XM_006227697.2 | XP_006227759.1 | 6 | 959 | |||
| XM_006227698.2 | XP_006227760.1 | 5 | 2621 | |||
| XM_006227699.2 | XP_006227761.1 | 5 | 519 | |||
| XM_008758644.1 | XP_008756866.1 | 6 | 1875 | |||

L3mbtl3
Genomic Map