siRNA Details
| Gene Symbol: L3mbtl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748666.3 | XP_003748714.1 | 8, 9 | 1141 | ||
| XM_006227693.2 | XP_006227755.1 | 7, 8 | 2797 | |||
| XM_006227695.2 | XP_006227757.1 | 8, 9 | 855 | |||
| XM_006227696.2 | XP_006227758.1 | 7, 8 | 774 | |||
| XM_006227697.2 | XP_006227759.1 | 7, 8 | 1060 | |||
| XM_006227698.2 | XP_006227760.1 | 6, 7 | 2722 | |||
| XM_006227699.2 | XP_006227761.1 | 6, 7 | 620 | |||
| XM_008758644.1 | XP_008756866.1 | 7, 8 | 1976 | |||

L3mbtl3
Genomic Map