siRNA Details
Gene Symbol: Epb41l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227709.1 | XP_006227771.1 | 18, 19 | 3033 | ||
XM_008758646.1 | XP_008756868.1 | 18, 19 | 3215 | |||
XM_008758647.1 | XP_008756869.1 | 18, 19 | 3331 | |||
XM_008758648.1 | XP_008756870.1 | 18, 19 | 2936 | |||
XM_008758649.1 | XP_008756871.1 | 19, 20 | 3171 | |||
XM_008758650.1 | XP_008756872.1 | 18, 19 | 6018 |
