siRNA Details
| Gene Symbol: Vars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213563.1 | NP_998728.1 | 7 | 1006 | ||
| XM_006255932.1 | XP_006255994.1 | 7 | 1059 | |||
| XM_006255933.1 | XP_006255995.1 | 7 | 604 | |||
| XM_006255934.1 | XP_006255996.1 | 7 | 1059 | |||

Vars2
Genomic Map