siRNA Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728921.4 | XP_002728967.2 | 14 | 2221 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uhrf1bp1| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728921.4 | XP_002728967.2 | 14 | 2221 | ||