siRNA Details
| Gene Symbol: Anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107613.3 | NP_001101083.1 | 20, 21 | 3089 | ||
| XM_006256193.2 | XP_006256255.1 | 21, 22 | 3164 | |||
| XM_006256195.2 | XP_006256257.1 | 20, 21 | 3101 | |||
| XM_006256196.2 | XP_006256258.1 | 21, 22 | 3164 | |||
| XM_006256197.2 | XP_006256259.1 | 20, 21 | 3101 | |||
| XM_008772813.1 | XP_008771035.1 | 21, 22 | 3164 | |||
| XM_008772814.1 | XP_008771036.1 | 13, 14 | 3025 | |||
| XM_008772815.1 | XP_008771037.1 | 11, 12 | 1602 | |||

Anks1a
Genomic Map