siRNA Details
| Gene Symbol: Cmtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014031.1 | NP_001014053.1 | 12 | 1330 | ||
| XM_008772816.1 | XP_008771038.1 | 12 | 1400 | |||
| GenBank mRNA | AY539880.1 | - | 1574 | |||
| BC085799.1 | - | 12 | 1330 | 7110279 | ||

Cmtr1
Genomic Map