siRNA Details
| Gene Symbol: Rrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772835.1 | XP_008771057.1 | 1, 2 | 132 | ||
| XM_228076.8 | XP_228076.4 | 1, 2 | 208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrp1b| Gene Symbol: Rrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772835.1 | XP_008771057.1 | 1, 2 | 132 | ||
| XM_228076.8 | XP_228076.4 | 1, 2 | 208 | |||