siRNA Details
| Gene Symbol: Dnmt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003964.1 | NP_001003964.1 | 10 | 964 | ||
| XM_006256250.2 | XP_006256312.1 | 9 | 987 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnmt3l| Gene Symbol: Dnmt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003964.1 | NP_001003964.1 | 10 | 964 | ||
| XM_006256250.2 | XP_006256312.1 | 9 | 987 | |||