siRNA Details
| Gene Symbol: Nepn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107632.1 | NP_001101102.1 | 2 | 417 | ||
| XM_006256491.2 | XP_006256553.1 | 2 | 702 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nepn| Gene Symbol: Nepn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107632.1 | NP_001101102.1 | 2 | 417 | ||
| XM_006256491.2 | XP_006256553.1 | 2 | 702 | |||