siRNA Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107633.1 | NP_001101103.1 | 6 | 2367 | ||
| XM_006256375.2 | XP_006256437.1 | 6 | 2453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcc2| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107633.1 | NP_001101103.1 | 6 | 2367 | ||
| XM_006256375.2 | XP_006256437.1 | 6 | 2453 | |||