siRNA Details
| Gene Symbol: Rev3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083966.1 | NP_001077435.1 | 29, 30 | 9108 | ||
| XM_006256524.1 | XP_006256586.1 | 29, 30 | 9113 | |||
| XM_006256525.2 | XP_006256587.1 | 30, 31 | 8871 | |||
| XM_008772987.1 | XP_008771209.1 | 29, 30 | 9174 | |||
| GenBank mRNA | BC086428.1 | - | 3, 4 | 339 | ||

Rev3l
Genomic Map