siRNA Details
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107636.1 | NP_001101106.1 | 20 | 2977 | ||
XM_006256449.2 | XP_006256511.1 | 21 | 3243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107636.1 | NP_001101106.1 | 20 | 2977 | ||
XM_006256449.2 | XP_006256511.1 | 21 | 3243 |