siRNA Details
Gene Symbol: Tmem161b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760644.1 | XP_008758866.1 | 2 | 272 |
Gene Symbol: LOC685655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770407.1 | XP_008768629.1 | 1 | 169 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175187, AK088393, BY239899, BC051663, AK035319.
