siRNA Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728994.4 | XP_002729040.2 | 6 | 1145 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
