siRNA Details
| Gene Symbol: Utp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107647.2 | NP_001101117.1 | 11 | 1437 | ||
| XM_006231836.2 | XP_006231898.1 | 11 | 1865 | |||
| XM_008760671.1 | XP_008758893.1 | 10 | 4060 | |||
| XM_008760672.1 | XP_008758894.1 | 11 | 3912 | |||
| XM_008760673.1 | XP_008758895.1 | 11 | 1669 | |||
| GenBank mRNA | BC107908.1 | - | 184 | |||
| BC131851.1 | - | 11 | 1437 | 7189311 | ||
| FQ211666.1 | - | 11 | 1488 | |||

Utp15
Genomic Map