siRNA Details
| Gene Symbol: Utp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107647.2 | NP_001101117.1 | 4 | 470 | ||
| XM_006231836.2 | XP_006231898.1 | 4 | 898 | |||
| XM_008760671.1 | XP_008758893.1 | 3 | 3093 | |||
| XM_008760672.1 | XP_008758894.1 | 4 | 2945 | |||
| XM_008760673.1 | XP_008758895.1 | 4 | 702 | |||
| GenBank mRNA | BC131851.1 | - | 4 | 470 | 7189311 | |
| FQ211666.1 | - | 4 | 521 | |||

Utp15
Genomic Map