siRNA Details
| Gene Symbol: Zfp366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231860.2 | XP_006231922.1 | 2 | 965 | ||
| XM_008760692.1 | XP_008758914.1 | 3 | 841 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp366| Gene Symbol: Zfp366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231860.2 | XP_006231922.1 | 2 | 965 | ||
| XM_008760692.1 | XP_008758914.1 | 3 | 841 | |||