siRNA Details
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168000.1 | NP_001161472.1 | 2, 3 | 279 | ||
XM_006231924.2 | XP_006231986.1 | 4, 5 | 374 | |||
XM_006231925.2 | XP_006231987.2 | 4, 5 | 781 | |||
GenBank mRNA | JQ013732.1 | - | 2, 3 | 1225 |
