siRNA Details
| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012077.1 | NP_001012077.1 | 13 | 1264 | ||
| XM_006232046.2 | XP_006232108.1 | 13 | 1355 | |||
| GenBank mRNA | BC089876.1 | - | 13 | 1264 | 7325487 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr12| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012077.1 | NP_001012077.1 | 13 | 1264 | ||
| XM_006232046.2 | XP_006232108.1 | 13 | 1355 | |||
| GenBank mRNA | BC089876.1 | - | 13 | 1264 | 7325487 | |