siRNA Details
| Gene Symbol: RGD1306502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107654.1 | NP_001101124.1 | 8 | 1154 | ||
| XM_006232047.2 | XP_006232109.1 | 8 | 1308 | |||
| XM_006232048.2 | XP_006232110.1 | 8 | 1096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306502| Gene Symbol: RGD1306502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107654.1 | NP_001101124.1 | 8 | 1154 | ||
| XM_006232047.2 | XP_006232109.1 | 8 | 1308 | |||
| XM_006232048.2 | XP_006232110.1 | 8 | 1096 | |||