siRNA Details
| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107657.1 | NP_001101127.1 | 17 | 2224 | ||
| XM_008760768.1 | XP_008758990.1 | 17 | 1967 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo10| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107657.1 | NP_001101127.1 | 17 | 2224 | ||
| XM_008760768.1 | XP_008758990.1 | 17 | 1967 | |||