siRNA Details
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749226.3 | XP_003749274.1 | 22 | 4275 | ||
XM_006232090.2 | XP_006232152.1 | 22 | 4275 | |||
XM_006232091.2 | XP_006232153.1 | 22 | 4150 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081302, CF539743, AB106872, BC067059.
Panther Classification:
Gene Ontology Categories:
