siRNA Details
Gene Symbol: Slc10a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025280.1 | NP_001020451.1 | 1 | 471 | ||
GenBank mRNA | DQ074435.1 | - | 1 | 471 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc10a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025280.1 | NP_001020451.1 | 1 | 471 | ||
GenBank mRNA | DQ074435.1 | - | 1 | 471 |