siRNA Details
| Gene Symbol: Lrrc31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232238.2 | XP_006232300.1 | 2 | 1172 | ||
| XM_006232240.2 | XP_006232302.1 | 4 | 901 | |||
| XM_008760950.1 | XP_008759172.1 | 5 | 3089 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc31| Gene Symbol: Lrrc31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232238.2 | XP_006232300.1 | 2 | 1172 | ||
| XM_006232240.2 | XP_006232302.1 | 4 | 901 | |||
| XM_008760950.1 | XP_008759172.1 | 5 | 3089 | |||