siRNA Details
| Gene Symbol: Samd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191703.1 | NP_001178632.1 | 4 | 564 | ||
| XM_006232225.2 | XP_006232287.1 | 5 | 1088 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd7| Gene Symbol: Samd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191703.1 | NP_001178632.1 | 4 | 564 | ||
| XM_006232225.2 | XP_006232287.1 | 5 | 1088 | |||