siRNA Details
| Gene Symbol: Arse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047885.1 | NP_001041350.1 | 5 | 718 | ||
| XM_006232280.2 | XP_006232342.1 | 6 | 905 | |||
| XM_006232281.2 | XP_006232343.1 | 6 | 910 | |||
| GenBank mRNA | BC168644.1 | - | 5 | 1020 | ||

Arse
Genomic Map