siRNA Details
| Gene Symbol: Plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107671.1 | NP_001101141.1 | 2 | 758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcxd3| Gene Symbol: Plcxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107671.1 | NP_001101141.1 | 2 | 758 | ||