siRNA Details
| Gene Symbol: Fbxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107672.1 | NP_001101142.1 | 4 | 1140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo4| Gene Symbol: Fbxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107672.1 | NP_001101142.1 | 4 | 1140 | ||