siRNA Details
Gene Symbol: Naa15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107674.1 | NP_001101144.1 | 16 | 2241 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Naa15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107674.1 | NP_001101144.1 | 16 | 2241 |