siRNA Details
| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004262.1 | NP_001004262.1 | 19 | 1901 | ||
| GenBank mRNA | BC079442.1 | - | 19 | 1901 | 7115484 | |
| FQ225755.1 | - | 1692 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog6| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004262.1 | NP_001004262.1 | 19 | 1901 | ||
| GenBank mRNA | BC079442.1 | - | 19 | 1901 | 7115484 | |
| FQ225755.1 | - | 1692 | ||||