siRNA Details
| Gene Symbol: Gpr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107677.1 | NP_001101147.1 | 1 | 927 | ||
| XM_006232401.2 | XP_006232463.1 | 2 | 1560 | |||
More Information
Other Species Targeted
Set Membership: |
|

Gpr87
Genomic Map