siRNA Details
| Gene Symbol: Igsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198768.2 | NP_942063.1 | 6 | 5735 | ||
| GenBank mRNA | AY273816.1 | - | 6 | 5779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf10| Gene Symbol: Igsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198768.2 | NP_942063.1 | 6 | 5735 | ||
| GenBank mRNA | AY273816.1 | - | 6 | 5779 | ||