siRNA Details
Gene Symbol: Plch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191707.1 | NP_001178636.1 | 14 | 1880 | ||
XM_006232442.2 | XP_006232504.1 | 16 | 3550 | |||
XM_006232446.2 | XP_006232508.1 | 15 | 2841 | |||
XM_008761049.1 | XP_008759271.1 | 15 | 2841 | |||
GenBank mRNA | FQ211591.1 | - | 15 | 2228 |
