siRNA Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191707.1 | NP_001178636.1 | 4 | 597 | ||
| XM_006232442.2 | XP_006232504.1 | 6 | 2264 | |||
| XM_006232446.2 | XP_006232508.1 | 5 | 1555 | |||
| XM_008761049.1 | XP_008759271.1 | 5 | 1555 | |||
| GenBank mRNA | FQ211591.1 | - | 5 | 942 | ||

Plch1
Genomic Map