siRNA Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191707.1 | NP_001178636.1 | 21 | 4634 | ||
| XM_006232442.2 | XP_006232504.1 | 24 | 6364 | |||
| XM_006232446.2 | XP_006232508.1 | 24 | 5678 | |||
| XM_008761049.1 | XP_008759271.1 | 23 | 5655 | |||
| GenBank mRNA | FQ211591.1 | - | 24 | 5066 | ||

Plch1
Genomic Map