siRNA Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749294.3 | XP_003749342.1 | 6 | 1997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tiparp| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749294.3 | XP_003749342.1 | 6 | 1997 | ||