siRNA Details
| Gene Symbol: B3galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013158.1 | NP_001013176.1 | 3 | 627 | ||
| XM_008761089.1 | XP_008759311.1 | 1 | 471 | |||
| GenBank mRNA | BC079206.1 | - | 3 | 627 | 7131789 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3galnt1| Gene Symbol: B3galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013158.1 | NP_001013176.1 | 3 | 627 | ||
| XM_008761089.1 | XP_008759311.1 | 1 | 471 | |||
| GenBank mRNA | BC079206.1 | - | 3 | 627 | 7131789 | |