siRNA Details
| Gene Symbol: Rbm46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135717.1 | NP_001129189.1 | 3 | 536 | ||
| XM_006232530.2 | XP_006232592.1 | 3 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm46| Gene Symbol: Rbm46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135717.1 | NP_001129189.1 | 3 | 536 | ||
| XM_006232530.2 | XP_006232592.1 | 3 | 531 | |||