siRNA Details
| Gene Symbol: Tlr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198769.2 | NP_942064.1 | 2 | 1500 | ||
| XM_008761102.1 | XP_008759324.1 | 3 | 1865 | |||
| GenBank mRNA | AY151255.1 | - | 2 | 1500 | ||
| AY151256.1 | - | 2 | 1500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlr2| Gene Symbol: Tlr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198769.2 | NP_942064.1 | 2 | 1500 | ||
| XM_008761102.1 | XP_008759324.1 | 3 | 1865 | |||
| GenBank mRNA | AY151255.1 | - | 2 | 1500 | ||
| AY151256.1 | - | 2 | 1500 | |||