siRNA Details
| Gene Symbol: Creb3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007093.1 | NP_001007094.1 | 2 | 254 | ||
| XM_006232627.2 | XP_006232689.1 | 3 | 312 | |||
| XM_008761147.1 | XP_008759369.1 | 2 | 409 | |||
| XM_008761148.1 | XP_008759370.1 | 2 | 484 | |||
| GenBank mRNA | AY750875.1 | - | 2 | 254 | ||
| AY750876.1 | - | 2 | 248 | |||

Creb3l4
Genomic Map