siRNA Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012078.2 | NP_001012078.2 | 6, 7 | 793 | ||
| XM_006232639.2 | XP_006232701.1 | 6, 7 | 673 | |||
| XM_006232640.2 | XP_006232702.1 | 6, 7 | 644 | |||
| XM_006232641.2 | XP_006232703.1 | 6, 7 | 873 | |||
| XM_008761150.1 | XP_008759372.1 | 6, 7 | 1367 | |||
| GenBank mRNA | BC081943.1 | - | 6, 7 | 737 | 7121310 | |

Sema4a
Genomic Map