siRNA Details
| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014038.2 | NP_001014060.2 | 6 | 938 | ||
| XM_006232870.2 | XP_006232932.1 | 6 | 941 | |||
| XM_006232871.2 | XP_006232933.1 | 6 | 2897 | |||
| GenBank mRNA | BC059161.1 | - | 6 | 894 | 6887770 | |

Tdrkh
Genomic Map