siRNA Details
| Gene Symbol: mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007696.1 | NP_001007697.1 | 1 | 142 | ||
| GenBank mRNA | BC082085.1 | - | 1 | 142 | 7133689 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpl9| Gene Symbol: mrpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007696.1 | NP_001007697.1 | 1 | 142 | ||
| GenBank mRNA | BC082085.1 | - | 1 | 142 | 7133689 | |